missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen PerOxide Solution, 3 wt% in H2O, Honeywell Fluka™
Contains ∼200 ppm acetanilide as stabilizer, 3 wt. % in H2O
52.60 € - 405.37 €
Chemical Identifiers
CAS | 7722-84-1 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
MDL Number | MFCD00011333 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15658360
|
Honeywell-Fluka
323381-25ML |
25 mL |
52.60 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15613820
|
Honeywell-Fluka
323381-500ML |
500 mL |
105.10 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15668360
|
Honeywell-Fluka
323381-4L |
4 L |
405.37 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
7722-84-1 | |
MFCD00011333 | |
3587191 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
23.3mmHg (30°C) | |
Hydrogen Peroxide Solution |
H2O2 | |
NONH for all modes of transport | |
1g/mL at 25°C | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01g/mol | |
1.1 (vs air) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title