missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
Boiling Point | 108.0°C |
Melting Point | -33.0°C |
Color | Yellow through Brown |
Physical Form | Powder |
Quantity | 25 L |
Linear Formula | H2O2 |
Fieser | 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145 |
Merck Index | 15, 4835 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether |
Formula Weight | 34 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction