Learn More
Hydrogen fluoride-pyridine, 65-70%, Thermo Scientific Chemicals
40.40 € - 106.53 €
Chemical Identifiers
CAS | 32001-55-1 |
---|---|
Molecular Formula | C5H6FN |
Molecular Weight (g/mol) | 99.11 |
MDL Number | MFCD00012436 |
InChI Key | GRJJQCWNZGRKAU-UHFFFAOYSA-N |
Synonym | pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine |
PubChem CID | 64774 |
IUPAC Name | pyridine;hydrofluoride |
SMILES | C1=CC=NC=C1.F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10317220
|
Thermo Scientific Acros
180070100 |
10 g | Plastic bottle |
40.40 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10773371
|
Thermo Scientific Acros
180070500 |
50 g | Plastic Bottle in Tin Can |
77.39 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10502401
|
Thermo Scientific
180071000 |
100 g | Plastic Bottle in Tin Can |
106.53 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
32001-55-1 | |
99.11 | |
GRJJQCWNZGRKAU-UHFFFAOYSA-N | |
64774 | |
C1=CC=NC=C1.F |
C5H6FN | |
MFCD00012436 | |
pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine | |
pyridine;hydrofluoride |
Specifications
32001-55-1 | |
1.1000g/mL | |
C5H6FN | |
16,286; 16,050; 05,538; 06,473; 09,399; 12,419 | |
pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine | |
C1=CC=NC=C1.F | |
99.11 | |
99.11 | |
Liquid |
Yellow to Brown | |
65-70% | |
MFCD00012436 | |
1.1 | |
GRJJQCWNZGRKAU-UHFFFAOYSA-N | |
pyridine;hydrofluoride | |
64774 | |
65-70% | |
Hydrogen fluoride-pyridine, 65-70% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 250-889-6
missing translation for 'tsca' : TSCA
- Hydrogen fluoride-pyridine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only