missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 5M, Pure, Bench Reagent, Fisher Chemical™

Product Code. 10695872
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH lt; 1
Color Colorless to Yellow
Packaging HDPE plastic bottle
Quantity 2.5 L
Physical Form Liquid
IUPAC Name chlorane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'