missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.1M, Honeywell Fluka™

Product Code. 15623540
Click to view available options
Quantity:
500 mL
2 L
4 L
4 x 4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.1 M HCl (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 0.1M
Density 1g/mL at 25°C
Vapor Pressure 190mmHg (25°C)
Quantity 500 mL
UN Number 1789
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric Acid Solution
compliance-icons
Product Identifier
  • Hydrochloric acid solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'