missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
ClH, CAS Number-7647-01-0, hydrogen chloride, hydrochloric acid, acide chlorhydrique, chlorwasserstoff, anhydrous hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
122.00 € - 188.00 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10510921
|
Fisher Chemical
J/4350/21 |
5 L | Plastic container |
122.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10439560
|
Fisher Chemical
J/4350/24 |
10 L | Bag in box |
135.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12616727
|
Fisher Chemical
J/4350/25 |
25 L | Plastic container |
188.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ClH, CAS Number-7647-01-0, hydrogen chloride, hydrochloric acid, acide chlorhydrique, chlorwasserstoff, anhydrous hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
2 | |
7647-01-0 | |
14mmHg @20°C | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Colorless | |
100°C | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrochloric acid, 0.1M, (0.1N)
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.