missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.02M (0.02N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
39.45 € - 86.40 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10733991
|
Fisher Chemical
J/4360/PB15 |
1 L |
39.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10538040
|
Fisher Chemical
J/4360/17 |
2.5 L |
86.40 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
2 | |
100°C | |
14&20 | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
7647-01-0 | |
HDPE plastic bottle | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 |
Safety and Handling
Product Identifier
- Hydrochloric acid, 0.02M, (0.02N)