missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, DILUT-IT Concentrate, 0.5N, J.T. Baker™

Product Code. 10761072
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Prepare fresh, reliable standard solutions quickly and accurately. Hydrochloric Acid, DILUT-IT Concentrate, 0.5N, J.T. Baker™ is a pre-mixed volumetric concentrate for dilution to 1L that is ideal for titrimetry and other forms of wet chemical analysis.

DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.

  • Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
  • Space saving: Products require only one-tenth of the space required for ready-made solutions
  • Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate

Specifications

Color Colorless
CAS 7647-01-0
Boiling Point 85°C
Concentration 0.5mol/L
Density 1.0g/cm3 at 20°C
Packaging LDPE ampule
Vapor Pressure 1.100 hPa (50°C)
Normality 0.5N
Quantity For dilution to 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.