missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate, For 1L standard solution, 0.5 M HCl (0.5N), Honeywell Fluka™

Product Code. 15615280
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 1L standard solution, 0.5 M HCl (0.5N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Boiling Point 100°C (literature)
CAS Min % ≥70.0000%, ≥20.0000%
Concentration 0.5 M HCl (0.5N)
CAS Max % <80.0000%, <30.0000%
Density 1.012 g/cm3
Packaging Ampoule
Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 1 Ea.
Linear Formula HCl
Physical Form Concentrate
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid concentrate, For 1L standard solution, 0.5 M HCl (0.5N)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'