Learn More
Hydrochloric acid, 4N solution in 1,4-Dioxane/water, Thermo Scientific Chemicals
73.06 € - 702.50 €
Specifications
Chemical Name or Material | Hydrochloric acid |
---|---|
Color | Colorless to Yellow |
Name Note | 4N Solution in 1, 4-dioxane/water |
CAS | 7647-01-0, 7732-18-5, 123-91-1 |
Concentration | 3.95 to 4.40N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10464272
|
Thermo Scientific Acros
366521000 |
100 mL | Glass bottle |
73.06 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10140902
|
Thermo Scientific Acros
366520025 |
2.5 L | Glass bottle |
702.50 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
4N Solution in 1, 4-dioxane/water | |
3.95 to 4.40N | |
Glass bottle | |
12°C | |
HCl | |
MFCD00011324 MFCD00792839 | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
313 | |
36.45 |
Colorless to Yellow | |
7647-01-0, 7732-18-5, 123-91-1 | |
1.1000g/mL | |
1.1 | |
ClH | |
Liquid | |
2924 | |
15, 4818 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
36.46 | |
CHEBI:17883 |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Hydrochloric acid, 4N solution in 1,4-Dioxane/water
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.