missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, purum p.a., Honeywell™
Purum p.a.,≥48%
31.13 € - 105.86 €
Chemical Identifiers
CAS | 10035-10-6 |
---|---|
Molecular Formula | BrH |
Molecular Weight (g/mol) | 80.91 |
MDL Number | MFCD00011323 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-N |
Synonym | hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide |
PubChem CID | 260 |
ChEBI | CHEBI:47266 |
IUPAC Name | bromane |
SMILES | Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15661690
|
Honeywell Research Chemicals
18730-250ML |
250 mL | N/A | N/A | N/A | |||||
15651690
|
Honeywell Research Chemicals
18730-1L |
1 L |
105.86 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
10035-10-6 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
bromane |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 | |
Br |
Specifications
10035-10-6 | |
8mmHg (25°C), 320psi (21.1°C) | |
MFCD00011323 | |
3587158 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
bromane | |
260 | |
80.91g/mol | |
purum p.a. |
1.49g/mL at 25°C | |
BrH | |
1788 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
Br | |
80.91 | |
CHEBI:47266 | |
≥48% | |
Hydrobromic Acid |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314-H335
missing translation for 'einecsNumber' : 233-113-0
missing translation for 'rtecsNumber' : MW3850000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrobromic Acid, purum p.a., Honeywell™