missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine monohydrobromide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 13775-80-9
Molecular Formula BrH5N2
Molecular Weight (g/mol) 112.958
MDL Number MFCD00065394
InChI Key DVHXJLRODLTJOD-UHFFFAOYSA-N
Synonym hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
PubChem CID 159693
IUPAC Name hydrazine;hydrobromide
SMILES NN.Br
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13775-80-9
112.958
DVHXJLRODLTJOD-UHFFFAOYSA-N
159693
NN.Br
BrH5N2
MFCD00065394
hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
hydrazine;hydrobromide

Specifications

≈190°C (dec.)
13775-80-9
MFCD00065394
hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
NN.Br
112.958
112.96
Hydrazine monohydrobromide
92° to 93°C
BrH5N2
UN2928
DVHXJLRODLTJOD-UHFFFAOYSA-N
hydrazine;hydrobromide
159693
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.