Learn More
Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals
50.75 € - 413.06 €
Chemical Identifiers
CAS | 7803-57-8 |
---|---|
Molecular Formula | H6N2O |
Molecular Weight (g/mol) | 50.06 |
MDL Number | MFCD00149931 |
InChI Key | IKDUDTNKRLTJSI-UHFFFAOYSA-N |
Synonym | hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 |
PubChem CID | 24654 |
ChEBI | CHEBI:35511 |
SMILES | O.NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367198
|
Thermo Scientific Alfa Aesar
016651.22 |
100 g |
50.75 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397198
|
Thermo Scientific Alfa Aesar
016651.36 |
500 g |
129.65 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377198
|
Thermo Scientific Alfa Aesar
016651.A3 |
2 kg |
244.22 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387198
|
Thermo Scientific Alfa Aesar
016651.57 |
4 x 500 g |
413.06 €
2000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states. It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7803-57-8 | |
50.06 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
24654 | |
O.NN |
H6N2O | |
MFCD00149931 | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
CHEBI:35511 |
Specifications
120°C to 121°C | |
7803-57-8 | |
Liquid | |
H6N2O | |
MFCD00149931 | |
75° (167°F) | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
hydrazine hydrate | |
24654 | |
50.06 (32.05 Anhydrous) | |
1.431 | |
Hydrazine Monohydrate |
-52°C | |
Colorless | |
>99% | |
H2NNH2·H2O | |
2030 | |
14,4771 | |
Soluble in chloroform,DMSO,methanol. Slightly soluble in water. | |
O.NN | |
50.06 | |
CHEBI:35511 | |
≥99% | |
1.027 |
Safety and Handling
GHS H Statement
H301-H310-H330-H350-H314-H318-H227-H317
P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H301+H311-H314-H317-H330-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDRAZINE AQUEOUS SOLUTION
missing translation for 'einecsNumber' : 206-114-9
missing translation for 'rtecsNumber' : MV4590000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.