Learn More
Hydrazine monohydrate, 98+%, Thermo Scientific Chemicals
24.00 € - 170.00 €
Chemical Identifiers
CAS | 7803-57-8 |
---|---|
Molecular Formula | H6N2O |
Molecular Weight (g/mol) | 50.06 |
MDL Number | MFCD00149931 |
InChI Key | IKDUDTNKRLTJSI-UHFFFAOYSA-N |
Synonym | hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 |
PubChem CID | 24654 |
ChEBI | CHEBI:35511 |
SMILES | O.NN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488880
|
Thermo Scientific Alfa Aesar
A14005.18 |
50 g |
24.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498880
|
Thermo Scientific Alfa Aesar
A14005.30 |
250 g |
30.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11447866
|
Thermo Scientific Alfa Aesar
A14005.36 |
500 g |
49.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457866
|
Thermo Scientific Alfa Aesar
A14005.0E |
2500 g |
170.00 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.
Solubility
Miscible with water and alcohol. Immiscible with chloroform, hydrocarbons and ether.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents, oxygen, copper, organic materials and zinc.
Chemical Identifiers
7803-57-8 | |
50.06 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
24654 | |
O.NN |
H6N2O | |
MFCD00149931 | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
CHEBI:35511 |
Specifications
120°C to 121°C | |
7803-57-8 | |
H2NNH2·H2O | |
UN2030 | |
14,4771 | |
Miscible with water and alcohol. Immiscible with chloroform,hydrocarbons and ether. | |
O.NN | |
50.06 | |
CHEBI:35511 | |
≥98% | |
1.431 | |
Hydrazine monohydrate |
-52°C | |
H6N2O | |
MFCD00149931 | |
75°C (167°F) | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
IKDUDTNKRLTJSI-UHFFFAOYSA-N | |
hydrazine hydrate | |
24654 | |
50.06 (32.05 Anhydrous) | |
Amine-like | |
1.027 |
Safety and Handling
GHS H Statement
H301-H310-H330-H350-H314-H318-H227-H317
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H301+H311-H314-H317-H330-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDRAZINE AQUEOUS SOLUTION
missing translation for 'einecsNumber' : 206-114-9
missing translation for 'rtecsNumber' : MV4590000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only