Learn More
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
32.90 € - 80.60 €
Chemical Identifiers
CAS | 10217-52-4, 7732-18-5 |
---|---|
Molecular Formula | H4N2 |
Molecular Weight (g/mol) | 32.05 |
MDL Number | MFCD00149931 |
InChI Key | OAKJQQAXSVQMHS-UHFFFAOYSA-N |
Synonym | hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 |
PubChem CID | 24654 |
ChEBI | CHEBI:35511 |
IUPAC Name | hydrazine;hydrate |
SMILES | NN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10513191
|
Thermo Scientific Acros
209592500 |
250 g | Glass bottle |
32.90 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10793445
|
Thermo Scientific Acros
209590010 |
1 kg | Glass bottle |
80.60 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
10217-52-4, 7732-18-5 | |
32.05 | |
OAKJQQAXSVQMHS-UHFFFAOYSA-N | |
24654 | |
hydrazine;hydrate |
H4N2 | |
MFCD00149931 | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
CHEBI:35511 | |
NN |
Specifications
117.2°C | |
10217-52-4, 7732-18-5 | |
Liquid | |
H2NNH2·xH2O | |
01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174 | |
14, 4771 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether | |
NN | |
32.05 | |
CHEBI:35511 | |
80% | |
1.028 | |
Hydrazine hydrate |
-57.0°C | |
Colorless | |
H4N2 | |
MFCD00149931 | |
91°C | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
OAKJQQAXSVQMHS-UHFFFAOYSA-N | |
hydrazine;hydrate | |
24654 | |
32.04 | |
1.4105 to 1.4145 | |
1.0280g/mL |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause cancer.
Very toxic to aquatic life with l
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plen
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : MV4590000
- Hydrazine hydrate, 80% (Hydrazine, 51%)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only