Learn More
Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals
20.90 € - 191.00 €
Chemical Identifiers
CAS | 10217-52-4, 7732-18-5 |
---|---|
Molecular Formula | H4N2 |
Molecular Weight (g/mol) | 32.05 |
MDL Number | MFCD00149931 |
InChI Key | OAKJQQAXSVQMHS-UHFFFAOYSA-N |
Synonym | hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 |
PubChem CID | 24654 |
ChEBI | CHEBI:35511 |
SMILES | NN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521794
|
Thermo Scientific Acros
296811000 |
100 mL | Glass bottle |
20.90 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348830
|
Thermo Scientific Acros
296815000 |
500 mL | Glass bottle |
48.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954151
|
Thermo Scientific Acros
296810025 |
2.5 L | Glass bottle |
191.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
10217-52-4, 7732-18-5 | |
32.05 | |
OAKJQQAXSVQMHS-UHFFFAOYSA-N | |
24654 | |
NN |
H4N2 | |
MFCD00149931 | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
CHEBI:35511 |
Specifications
109.4°C | |
10217-52-4, 7732-18-5 | |
55% | |
H2NNH2·xH2O | |
01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174 | |
14,4771 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether | |
NN | |
32.05 | |
CHEBI:35511 | |
55% (Hydrazine,35%) | |
1.023 | |
Hydrazine hydrate, 55% (Hydrazine 35%) |
-65.0°C | |
Liquid | |
H4N2 | |
MFCD00149931 | |
>100°C | |
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739 | |
OAKJQQAXSVQMHS-UHFFFAOYSA-N | |
hydrazine;hydrate | |
24654 | |
32.04 | |
1.3870 to 1.3910 | |
1.0230g/mL |
Safety and Handling
GHS H Statement
Fatal if inhaled.
May cause cancer.
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Causes severe skin b
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : MV4590000
- Hydrazine hydrate, 55% (Hydrazine, 35%)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Hydrazine (hydrate),Hydrazine
RUO – Research Use Only