missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Tetracycline Screened Fetal Bovine Serum
Brand: Cytiva SH30070.03TH
This item is not returnable.
View return policy
Description
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Tetracycline Screened | |
Sterile | |
Defined, Specialty screened | |
500 mL | |
No | |
No |
United States | |
500 mL | |
Sequential-filtered, 40 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
less than or equal to 10 EU/mL | |
less than or equal to 10 EU/mL |