Learn More
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Gamma Irradiated Characterized Fetal Bovine Serum (Australia)
Brand:  Cytiva (Formerly GE Healthcare Life Sciences) SH30084.04IR
Product Code. 12694197
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality
Fetal Bovine Serum
Sequential Triple-filtered, 100 nm, True Pool Technology
<–1°C
1000 mL
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus