missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ HyClone™ Gamma Irradiated Characterized Australian Fetal Bovine Serum
Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.
Brand: Cytiva Hyclone™ SH30084.03IR
This item is not returnable.
View return policy
Description
- Filtered through 3 sequential 0.1µm pore size filters
- Provided in a 500mL bottle
Specifications
Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Characterized | |
500 mL | |
Sequential Triple-filtered, 100 nm, True Pool Technology | |
Gamma Irradiated | |
USDA Inspected |
USDA Regulations; AQIS | |
Australia | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction