missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Tetracyclin Screened
Maximize cell culture growth. HyClone™ Fetal Bovine Serum (U.S.), Tetracyclin Screened contains undetectable tetracycline levels and is designed for researchers using tetracycline-regulated gene expression in cultured cells, and filtered through sequential 40nm pore-size filters.
Brand: Cytiva SH30070.03T
This item is not returnable.
View return policy
Description
- Filtered through sequential 40nm (0.04μm) pore-size rated filters
- Further heat inactivation or irradiation processing can be done upon request
Specifications
1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 40 nm | |
None | |
≤ 10 EU/mL | |
≤ 10 EU/mL | |
USDA Inspected |
EMEA Certified | |
Fetal Bovine Serum (U.S.), Tetracycline Screened | |
Tetracycline Screened | |
Sterile | |
Defined, Specialty screened | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction