missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Embryonic Stem (ES) Cell Screened
Augment growth of cells with U.S.-sourced HyClone™ Embryonic Stem (ES) Cell Screened Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.
226.00 € - 956.00 €
Specifications
Additional Assays | 1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C |
---|---|
Certifications/Compliance | EMEA Certified |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Country of Origin | United States |
Specialty Type | ES cell screened |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
12339792
|
Cytiva
SH30070.03EH |
500 mL | Heat Inactivated, ES Screened | N/A | ||||||
11501831
|
Cytiva
SH30070.02E |
100 mL | Embryonic Stem Cell Screened | N/A | ||||||
10117272
|
Cytiva
SH30070.03E |
500 mL | Embryonic Stem Cell Screened | N/A | ||||||
Description
- Filtered through sequential 40 nm (0.04μm) pore-size rated filters
- Endotoxin: ≤ 10 EU/mL; Hemoglobin ≤ 10mg/dL
Specifications
1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
ES cell screened | |
Sequential-filtered, 40 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 10 mg/dL |
EMEA Certified | |
United States | |
Sterile | |
Defined, Specialty screened | |
≤ 10 EU/mL | |
USDA Inspected |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Embryonic Stem (ES) Cell Screened