missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin
Sterile-filtered and quality-tested to ensure dependable, reproducible cell growth
Brand: Cytiva SH30084.50M
This item is not returnable.
View return policy
Description
- Meets the requirements of most users
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Certificate of Suitability No. R1-CEP 2000-384 | |
Serum | |
50 mL | |
Characterized | |
≤ 25 EU/mL | |
HyClone™ |
Australia | |
Fetal Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
Mesenchymal Stem Cell Screened | |
≤ 25 mg/dL |