Learn More
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
29.70 € - 396.00 €
Chemical Identifiers
CAS | 68131-04-4 |
---|---|
Molecular Formula | C9H8Na2O4 |
Molecular Weight (g/mol) | 226.139 |
MDL Number | MFCD00135560 |
InChI Key | HMGUIQPKFUZDPV-UHFFFAOYSA-L |
Synonym | humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt |
PubChem CID | 21908873 |
IUPAC Name | disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate |
SMILES | C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10284820
|
Thermo Scientific Acros
120860050 |
5 g | Glass bottle |
29.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102920
|
Thermo Scientific Acros
120861000 |
100 g | Plastic bottle |
66.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10560211
|
Thermo Scientific Acros
120860010 |
1 kg | Plastic bottle |
396.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68131-04-4 | |
226.139 | |
HMGUIQPKFUZDPV-UHFFFAOYSA-L | |
21908873 | |
C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
C9H8Na2O4 | |
MFCD00135560 | |
humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt | |
disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate |
Specifications
68131-04-4 | |
Black | |
C9H8Na2O4 | |
humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt | |
C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
226.139 | |
45 to 70% | |
Authentic | |
Crystalline Powder |
>300.0°C | |
45-70% | |
MFCD00135560 | |
HMGUIQPKFUZDPV-UHFFFAOYSA-L | |
disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate | |
21908873 | |
Technical | |
14% to 25% (105°C, 4 hrs) | |
Humic acid, sodium salt, tech., 50-60% (as humic acid) |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 268-608-0
missing translation for 'rtecsNumber' : MT6550000
missing translation for 'tsca' : TSCA
RUO – Research Use Only