missing translation for 'onlineSavingsMsg'
Learn More

Humic acid, Thermo Scientific Chemicals

50.75 € - 193.97 €

Chemical Identifiers

CAS 1415-93-6
MDL Number MFCD00147177
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388898
SDS View Documents
Thermo Scientific Alfa Aesar
041747.06
5 g
50.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
11398898
SDS View Documents
Thermo Scientific Alfa Aesar
041747.14
25 g
193.97 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1415-93-6
MFCD00147177

Specifications

1415-93-6
Slightly soluble in water,with much swellingSoluble in organic solvents like acetone and benzenol. Slightly soluble in water.
Humic acid
MFCD00147177
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-809-6

missing translation for 'rtecsNumber' : MT6544000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Humic acid, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.