missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human Sonic Hedgehog (SHH) Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
106.00 € - 792.00 €
Specifications
Gene ID (Entrez) | 6469 |
---|---|
Molecular Weight (g/mol) | 20 to 24 kDa |
Species | Human |
Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801426
|
Proteintech™
HZ-1306-5UG |
5 μg |
106.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16891416
|
Proteintech™
HZ-1306-10UG |
10 μg |
182.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881416
|
Proteintech™
HZ-1306-100UG |
100 μg |
792.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
6469 | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 90% |
20 to 24 kDa | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically ≤ 350 ng/mL EC50 | |
Lyophilized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title