missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GATA-4 ELISA Kit
Human GATA-4 quantitates human GATA-4 in serum, plasma, supernatant.
Brand: Invitrogen™ EH211RBX5
This item is not returnable.
View return policy
Description
Human GATA-4 quantitates human GATA-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GATA-4.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.Specifications
ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
0.85 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
GATA4 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.85 to 200 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P43694 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
GATA4 | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction