missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Alpha-Galactosidase A/GLA ELISA Kit
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant.
654.00 € - 4995.00 €
Specifications
Accession Number | Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase |
---|---|
Assay Range | 45 to 10,000 pg/mL |
Assay Sensitivity | 45 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17134979
|
Invitrogen™
EH21RBX5 |
5 x 96 Tests |
2635.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
17804100
|
Invitrogen™
EH21RBX10 |
10 x 96 Tests |
4995.00 €
Pack of 10 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17184503
|
Invitrogen™
EH21RB |
96 Tests |
654.00 €
Pack of 96 |
In Stock
Log in to see stock. |
|||||
Description
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Alpha-Galactosidase A/GLA.
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.Specifications
Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase | |
45 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
GLA | |
<10% | |
HRP | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
alpha Galactosidase | |
4 hr 45 min |
45 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P06280 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title