missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10211-1-AP-20UL
This item is not returnable.
View return policy
Description
HspBP1 is an Hsp70-interacting protein that was first isolated from a human heart cDNA library using the yeast two-hybrid system (1,2). The ATPase domain of Hsp70 binds HspBP1 and upon binding, inhibits Hsp70 chaperone activity. More recently HspBP1 was found to promote nucleotide dissociation from Hsc70 (3) and is capable of altering the confirmation of the Hsp70 ATPase domain (4). HspBP1 is capable of regulating any Hsp70 activity that requires ATPase.Specifications
HSPBP1 | |
Polyclonal | |
Unconjugated | |
HSPBP1 | |
FES1, Hsp70 binding protein 1, Hsp70 binding protein 2, Hsp70 interacting protein 1, Hsp70 interacting protein 2, HSPBP, HSPBP1, HspBP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23640, 246146, 66245 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6IMX7, Q99P31, Q9NZL4 | |
Hspbp1 | |
HSPBP1 Fusion Protein Ag0282 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction