Learn More
HSP90AB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11405-1-AP-20UL
 View more versions of this product
Product Code. 16828463
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
Specifications
Specifications
HSP90AB1
Polyclonal
Unconjugated
HSP90AB1
D6S182, Heat shock 84 kDa, Heat shock protein HSP 90 beta, HS90B, HSP 84, HSP 90, HSP84, HSP90 BETA, HSP90AB1, HSP90B, HSPC2, HSPCB
Rabbit
Antigen Affinity Chromatography
RUO
15516, 301252, 3326
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08238, P11499, P34058
HSP90AB1
HSP90AB1 Fusion Protein Ag1947
20 μL
Primary
Rat, Mouse, Human