missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90AB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11405-1-AP-150UL
This item is not returnable.
View return policy
Description
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.Specifications
HSP90AB1 | |
Polyclonal | |
Unconjugated | |
HSP90AB1 | |
D6S182, Heat shock 84 kDa, Heat shock protein HSP 90 beta, HS90B, HSP 84, HSP 90, HSP84, HSP90 BETA, HSP90AB1, HSP90B, HSPC2, HSPCB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15516, 301252, 3326 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08238, P11499, P34058 | |
HSP90AB1 | |
HSP90AB1 Fusion Protein Ag1947 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |