Learn More
HSP90 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13171-1-AP-150UL
 View more versions of this product
Product Code. 16891244
  • 410.00 € / 150µL
Explore more special offers
Description
Description
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).
Specifications
Specifications
HSP90
Polyclonal
Unconjugated
HSP90AA1
Heat shock 86 kDa, HSP 86, HSP86, Hsp89, HSP89A, HSP90, HSP90A, HSP90AA1, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, LAP2
Rabbit
Antigen Affinity Chromatography
RUO
15519, 299331, 3320
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07900, P07901, P82995
Hsp90aa1
HSP90 Fusion Protein Ag3826
150 μL
Primary
Human, Mouse, Rat