missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Howard Leight™ Leightning™ L2 Ear Muffs
Click to view available options
Howard Leight™ Leightning L2 Ear Muffs; Model: L2F; Noise Reduction Rating (NRR): 27 dB
Description
- EN 352-1
- Suitable for folding for fast-paced environments
- Air Flow Control technology delivers clarity
- Snap in ear cushions for maximum wearer comfort and efficiency
- Telescopic heigh adjustment for good fit
- High degree of comfort for most tasks
Specifications
Specifications
Color | Black |
Certifications/Compliance | ANSI S3.19-1974 |
For Use With (Application) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel |
Model | L2F |
Noise Reduction Rating (NRR) | 27 dB |
Product Type | Ear Muffs |
Material | Steel |
Style | Over-the-head |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction