missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian Protection™ RAE8815 Thermal Protection Gloves
missing translation for 'orderingAttributeHoverText'
Size:
1
Description
- Suitable for handling warm pieces in mechanical or glass industries.
- 100% brushed cotton inside and out to provide thermal insulation and comfort.
- Cotton lining for comfort and sweat absorption.
- Straight thumb.
- Canvas cuff.
Spécification
Spécification
Certifications/Compliance | CE CATEGORY II Intermediary risks. EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/06/12/0490, EC Attestation Number 0075/797/162/06/12/0490 |
Classification | Dexterity: 5 |
Cuff Material | Canvas |
Cut Resistance Rating | 2 |
Exterior Finish | Brushed Cotton |
Material | Cotton |
Product Type | Gloves |
Size | 1 |
Temperature (Metric) Max | 200°C |
Lining | Brushed Cotton Drill |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'