Learn More
Homomorpholine hydrochloride, 98%, Thermo Scientific Chemicals
209.04 € - 812.04 €
Chemical Identifiers
CAS | 178312-62-4 |
---|---|
Molecular Formula | C5H12ClNO |
Molecular Weight (g/mol) | 137.607 |
MDL Number | MFCD02683070 |
InChI Key | LJRCWNIWOVZLKS-UHFFFAOYSA-N |
Synonym | 1,4-oxazepane hydrochloride, homomorpholine hydrochloride, 1,4-oxazepine, hexahydro-, hydrochloride, homomorpholine hcl, 1,4-oxazepan hydrochloride, 1,4 oxazepane hydrochloride, hexahydro-1,4-oxazepine hydrochloride, homomorpholine-hydrochloride, homo-morpholine hydrochloride, homomorph-oline hydrochloride |
PubChem CID | 23090214 |
IUPAC Name | 1,4-oxazepane;hydrochloride |
SMILES | C1CNCCOC1.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460367
|
Thermo Scientific Alfa Aesar
B21749.03 |
1 g |
209.04 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470367
|
Thermo Scientific Alfa Aesar
B21749.06 |
5 g |
812.04 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Homomorpholine Hydrochloride is a useful synthetic intermediate. It is used to prepare homomorpholine oxazolidinone with antibacterial activities. It is also used to synthesize A2A adenosine receptor antagonists for the treatment of Parkinson's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySlightly soluble in water.
Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
178312-62-4 | |
137.607 | |
LJRCWNIWOVZLKS-UHFFFAOYSA-N | |
23090214 | |
C1CNCCOC1.Cl |
C5H12ClNO | |
MFCD02683070 | |
1,4-oxazepane hydrochloride, homomorpholine hydrochloride, 1,4-oxazepine, hexahydro-, hydrochloride, homomorpholine hcl, 1,4-oxazepan hydrochloride, 1,4 oxazepane hydrochloride, hexahydro-1,4-oxazepine hydrochloride, homomorpholine-hydrochloride, homo-morpholine hydrochloride, homomorph-oline hydrochloride | |
1,4-oxazepane;hydrochloride |
Specifications
178312-62-4 | |
C5H12ClNO | |
Hygroscopic | |
Slightly soluble in water. | |
C1CNCCOC1.Cl | |
137.607 | |
137.61 | |
Homomorpholine hydrochloride |
158°C to 164°C | |
MFCD02683070 | |
1,4-oxazepane hydrochloride, homomorpholine hydrochloride, 1,4-oxazepine, hexahydro-, hydrochloride, homomorpholine hcl, 1,4-oxazepan hydrochloride, 1,4 oxazepane hydrochloride, hexahydro-1,4-oxazepine hydrochloride, homomorpholine-hydrochloride, homo-morpholine hydrochloride, homomorph-oline hydrochloride | |
LJRCWNIWOVZLKS-UHFFFAOYSA-N | |
1,4-oxazepane;hydrochloride | |
23090214 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.