missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Space Saver Plate for SE 235 Multiple Gel Caster
Space Saver Plate for SE 235 Multiple Gel Caster
Brand: Hoefer™ SE237
This item is not returnable.
View return policy
Description
- For use with SE 235 Multiple Gel Caster
- Material: Acrylic plastic
- Used to fill up space in caster where less than maximum number of gels are being cast
- 6 mm thick
Specifications
SE 235 Multiple Gel Caster | |
Acrylic | |
0.6 cm |
Space Saver Plate | |
0.23 in. |