Learn More
HA Tag Polyclonal Antibody (SG77), Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 715500
397.10 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
HA Tag Polyclonal antibody specifically detects HA Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
HA Tag | |
SG77 | |
Unconjugated | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
A synthetic peptide corresponding to the nine amino acid sequence HA-tag (YPYDVPDYA). | |
100 μg | |
Primary | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
-20°C | |
IgG |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AA399745 | Certificate of Analysis | 30/01/2025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.