Learn More
HLA class I ABC Mouse anti-Human, CoraLite 488, Clone: 5C5B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66013-100UL
Product Code. 16800696
  • 450.00 € / 100µL
Explore more special offers
Description
Description
The HLA class I ABC antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human HLA class I ABC. This antibody recognizes human, pig antigen. The HLA class I ABC antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Specifications
Specifications
HLA class I ABC
Monoclonal
CoraLite 488
100 μL
3105
IgG2b
Immunofluorescence, Flow Cytometry
5C5B7
Mouse
Primary
Human, Pig