Learn More
Histamine, Thermo Scientific Chemicals
85.02 €
Chemical Identifiers
CAS | 51-45-6 |
---|---|
Molecular Formula | C5H9N3 |
Molecular Weight (g/mol) | 111.15 |
MDL Number | MFCD00005210,MFCD00128939 |
InChI Key | NTYJJOPFIAHURM-UHFFFAOYSA-N |
Synonym | histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine |
PubChem CID | 774 |
ChEBI | CHEBI:18295 |
SMILES | NCCC1=CN=CN1 |
Description
Histamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHistamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.
Solubility
Soluble in water, alcohol, and hot chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Hygroscopic. Handle and store under inert gas. Air sensitive.
Chemical Identifiers
51-45-6 | |
111.15 | |
NTYJJOPFIAHURM-UHFFFAOYSA-N | |
774 | |
NCCC1=CN=CN1 |
C5H9N3 | |
MFCD00005210,MFCD00128939 | |
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
CHEBI:18295 |
Specifications
51-45-6 | |
C5H9N3 | |
UN2811 | |
14,4719 | |
Soluble in water,alcohol,and hot chloroform. | |
NCCC1=CN=CN1 | |
111.15 | |
CHEBI:18295 | |
Powder |
White | |
MFCD00005210,MFCD00128939 | |
2012 | |
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
NTYJJOPFIAHURM-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethanamine | |
774 | |
111.15 | |
Histamine |
Safety and Handling
GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
H301-H315-H317-H319-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-100-6
missing translation for 'rtecsNumber' : MS1050000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Histamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1H-Imidazole-4-ethanamine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.