missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Cobalt Spin Columns, 0.2 mL

Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.

Brand:  Thermo Scientific™ 89967

181.05 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.



Product Code. 11844181

  • 255.00 € / Pack of 25
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.

Additional Information and Online Ordering

HisPur Cobalt Resins, Columns and Kits use a tetradentate chelating resin charged with divalent cobalt (Co2+)to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types. HisPur Cobalt Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high throughput needs. With these options, high-quality His-tag protein purification is possible at nearly any scale. HisPur Cobalt Resin is superior to traditional nickel-based immobilized metal affinity chromatography (IMAC) systems. Compared to Ni-IDA and tetradentate Ni2+chelate resins, HisPur Resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole). Use HisPur Columns and Cartridges to obtain high-purity His-tagged proteins with no metal contamination.

Highlights:

  • High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions

  • Low metal leaching – no metal contamination in eluted histidine-tagged protein sample

  • Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives

  • Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates

  • Cost effective – resin can be reused several times or discarded after single use

  • Superior – performs better than other commercially available IMAC Resins

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
  • Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin

Includes:

Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.2 mL
Store at 4°C
HisPur™
Recombinant
Spin Column
Microcentrifuge, Spin Column
HisPur Cobalt Spin Columns, 0.2mL
Liquid Suspension
25 Columns
His-Tagged Protein
Metal Chelate, Divalent Cobalt
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HisPur Cobalt Spin Columns, 0.2 mL, 25 Columns

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.