Learn More
Scientific Industries SI™ High-density Foam Inserts
Holds vessels in place while mixing. Scientific Industries SI™ High-density Foam Inserts fit snugly inside the Vortex-Genie 2 and Vortex-Genie Pulse Recessed Platform.
Brand: Scientific Industries SI™ 503-0283-00
This item is not returnable.
View return policy
Description
- For use with Vortex-Genie 2 and Vortex-Genie Pulse Recessed Platform
- Accommodates:
- 9-16mm Tubes
- 14-29mm Tubes
- 29-37mm Tubes
- Blank Foam Insert provides customization for oddly-shaped vessels
- All items included in Large Sample Set (Modle 302)
Specifications
Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse for beakers, flasks and large tubes. | |
An assortment of Foam Inserts for tubes of various diameters (9mm to 37mm), 1 Blank Foam Insert for customization. | |
Recessed Platform 15547325 |