Learn More
Hexanes, mixed isomers, Environmental Grade, Thermo Scientific Chemicals
Description
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. It is also involved in the preparation of organolithiums compounds namely butyl lithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.67 g/mL |
Boiling Point | 65°C to 70°C |
Quantity | 4 L |
Flash Point | <−23°C (−9°F) |
Physical Form | Liquid |
Refractive Index | 1.3795 |
UN Number | UN1208 |
Solubility Information | Miscible with alcohol,chloroform,acetone and ether. Immiscible with water. |
Show More |
RUO – Research Use Only