missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, For residue analysis, ≥99.0%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
Melting Point | -95°C (literature) |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Density | 0.659 g/mL at 25 °C(lit.) |
Boiling Point | 66°C to 69°C |
Quantity | 1 L |
Flash Point | −26°C |
Packaging | Glass Bottle |
Linear Formula | CH3(CH2)4CH3 |
UN Number | UN1208 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'