missing translation for 'onlineSavingsMsg'
Learn More

Hexamine, Extra Pure, SLR, Fisher Chemical™

Product Code. 10305900
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Hexamine, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
MDL Number 6895
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Color White
pH 8.5 to 9.5
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 140.19g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.