missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals
11.40 € - 181.91 €
Chemical Identifiers
CAS | 107-46-0 |
---|---|
InChI Key | UQEAIHBTYFGYIE-UHFFFAOYSA-N |
Synonym | hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide |
PubChem CID | 24764 |
ChEBI | CHEBI:78002 |
IUPAC Name | trimethyl(trimethylsilyloxy)silane |
SMILES | C[Si](C)(C)O[Si](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10523371
|
Thermo Scientific Acros
194790100 |
10 mL | Glass bottle | N/A | |||||
10276870
|
Thermo Scientific Acros
194791000 |
100 mL | Glass bottle |
31.66 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10114120
|
Thermo Scientific Acros
194795000 |
500 mL | Glass bottle |
55.38 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11945811
|
Thermo Scientific Acros
194790025 |
2.5 L | Glass bottle |
181.91 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentChemical Identifiers
107-46-0 | |
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide | |
CHEBI:78002 | |
C[Si](C)(C)O[Si](C)(C)C |
UQEAIHBTYFGYIE-UHFFFAOYSA-N | |
24764 | |
trimethyl(trimethylsilyloxy)silane |
Specifications
107-46-0 | |
0.7600g/mL | |
98+% | |
(CH3)3SiOSi(CH3)3 | |
08,240; 09,55; 10,196 | |
UQEAIHBTYFGYIE-UHFFFAOYSA-N | |
trimethyl(trimethylsilyloxy)silane | |
24764 | |
162.38 | |
Liquid |
Colorless | |
Authentic | |
C6H18OSi2 | |
1.3765 to 1.3785 (20°C, 589nm) | |
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide | |
C[Si](C)(C)O[Si](C)(C)C | |
162.379 | |
CHEBI:78002 | |
≥98.0% (GC) | |
Hexamethyldisiloxane |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title