Learn More
Hexaamminecobalt(III) chloride, 97%, Thermo Scientific Chemicals
76.60 € - 229.00 €
Chemical Identifiers
CAS | 10534-89-1 |
---|---|
Molecular Formula | [Co(NH3)6]Cl3 |
MDL Number | MFCD00036304 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371309
|
Thermo Scientific Alfa Aesar
013769.14 |
25 g |
76.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361309
|
Thermo Scientific Alfa Aesar
013769.22 |
100 g |
229.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hexaamminecobalt(III) chloride is used in X-ray crystallography and NMR. It is useful in DNA condensation studies. It is suitable for induction of transitions of DNA from B to A or Z forms, induction of nucleic acid crystal growth, stabilization of tertiary tRNA interactions, preparation of ligation buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10534-89-1 | |
[Co(NH3)6]Cl3 | |
MFCD00036304 |
Specifications
10534-89-1 | |
Crystalline | |
[Co(NH3)6]Cl3 | |
Soluble in ammonia. | |
Odorless | |
1.71 g/cm3 |
Orange | |
97% | |
MFCD00036304 | |
267.48 | |
Orange powder | |
Hexaamminecobalt(III) chloride |
Safety and Handling
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350
missing translation for 'einecsNumber' : 234-103-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only