missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezers

Protect samples for −10° to −40°C applications without compromise using Thermo Scientific™ HERAfreeze™ HFU B Series −40°C Upright Ultra-Low Temperature Freezers

Specifications

Type Ultra-Low Freezer, Upright
Temperature Range -10°C to -40°C
Door Style Outer (1), Inner (5)
No. of Doors 1
Inner Doors 5
View More Specs

Description

Description

Protect samples for -10° to -40°C applications without compromise using Thermo Scientific™ HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezers. Designed for daily sample protection and dependability, these freezers feature microprocessor control and three capacities.

Features:
  • Knee-level, easy-to-use microprocessor control panel
  • Optional CO2 or LN2 safety back-up system for additional protection in the event of a power or mechanical failure
  • Vacuum relief port permits easy access after door openings
  • Power Management System with low voltage surge protection and buck/boost
  • Easy-to-remove air filter
  • Single 1 in. (25mm) access port
Construction/Design
  • Painted galveneel interior (stainless steel option available) in three sizes: 13, 17, and 23 cu. ft. (240, 320, 400L) to accommodate specific lab space and capacity requirements
  • Five interior compartment doors to reduce cold air loss, and to improve temperature recovery after door closings (five door option available)
  • Heavy gauge, cold-rolled steel exterior construction with a powder coat paint finish that resists chipping and rust
  • 5 in. (127mm) foamed-in-place, polyurethane insulation
  • Sturdy, solid, stainless-steel shelving to support a full product load
  • Ergonomic, lockable door handle
  • Water-cooled condenser option available

Warranty varies by country, please contact us for details

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.