Learn More
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
51.20 € - 511.55 €
Chemical Identifiers
CAS | 336-59-4 |
---|---|
Molecular Formula | C8F14O3 |
Molecular Weight (g/mol) | 410.06 |
MDL Number | MFCD00000432 |
InChI Key | UFFSXJKVKBQEHC-UHFFFAOYSA-N |
Synonym | heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization |
PubChem CID | 67643 |
ChEBI | CHEBI:39424 |
IUPAC Name | 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate |
SMILES | FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479622
|
Thermo Scientific Alfa Aesar
A10744.06 |
5 g | N/A | ||||||
11489622
|
Thermo Scientific Alfa Aesar
A10744.14 |
25 g | N/A | ||||||
11499622
|
Thermo Scientific Alfa Aesar
A10744.22 |
100 g |
511.55 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.
Solubility
Miscible with halogenated solvents. Immiscible with polar solvents.
Notes
Incompatible with moist air, water, strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
336-59-4 | |
410.06 | |
UFFSXJKVKBQEHC-UHFFFAOYSA-N | |
67643 | |
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate |
C8F14O3 | |
MFCD00000432 | |
heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization | |
CHEBI:39424 | |
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F |
Specifications
336-59-4 | |
1.662 | |
Stench | |
(CF3CF2CF2CO)2O | |
MFCD00000432 | |
Moisture sensitive | |
Miscible with halogenated solvents. Immiscible with polar solvents. | |
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F | |
410.06 | |
CHEBI:39424 | |
98% |
-43°C | |
108°C to 110°C | |
C8F14O3 | |
1.287 | |
UN3265 | |
heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization | |
UFFSXJKVKBQEHC-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate | |
67643 | |
410.06 | |
Heptafluorobutyric anhydride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 206-410-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Heptafluorobutyric anhydride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Heptafluorobutyric anhydride
RUO – Research Use Only