Learn More
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
60.70 € - 681.39 €
Chemical Identifiers
CAS | 375-22-4 |
---|---|
Molecular Formula | C4HF7O2 |
Molecular Weight (g/mol) | 214.039 |
MDL Number | MFCD00004171 |
InChI Key | YPJUNDFVDDCYIH-UHFFFAOYSA-N |
Synonym | heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech |
PubChem CID | 9777 |
ChEBI | CHEBI:39426 |
IUPAC Name | 2,2,3,3,4,4,4-heptafluorobutanoic acid |
SMILES | C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11422717
|
Thermo Scientific Alfa Aesar
A14907.14 |
25 g |
60.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432717
|
Thermo Scientific Alfa Aesar
A14907.22 |
100 g |
173.87 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442717
|
Thermo Scientific Alfa Aesar
A14907.36 |
500 g |
681.39 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
Solubility
Miscible with water, toluene, ether, ethanol and benzene.
Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..
Chemical Identifiers
375-22-4 | |
214.039 | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
9777 | |
2,2,3,3,4,4,4-heptafluorobutanoic acid |
C4HF7O2 | |
MFCD00004171 | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
CHEBI:39426 | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O |
Specifications
375-22-4 | |
1.648 | |
None | |
C4HF7O2 | |
1.3 | |
UN3265 | |
Light sensitive | |
Miscible with water,toluene,ether,ethanol and benzene. | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O | |
214.039 | |
CHEBI:39426 | |
99% |
-18°C | |
120°C to 121°C | |
Unpleasant | |
CF3CF2CF2CO2H | |
MFCD00004171 | |
1426882 | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutanoic acid | |
9777 | |
214.04 | |
Heptafluorobutyric acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I
missing translation for 'einecsNumber' : 206-786-3
missing translation for 'rtecsNumber' : ET4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Heptafluorobutyric acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Heptafluorobutyric acid
RUO – Research Use Only