missing translation for 'onlineSavingsMsg'
Learn More

HEPES, free acid, >98%, MP Biomedicals™

Product Code. 11344389
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color White
Assay Percent Range >98%
Quantity 1 kg
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Formula Weight 238.3
Physical Form Powder
Chemical Name or Material HEPES
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.