missing translation for 'onlineSavingsMsg'
Learn More

HEPES, ≥99% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15865158
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Melting Point ∼236°C (decomposition)
Physical Form Crystalline Powder
Quantity 250 g
Packaging Plastic bottle
Beilstein 883043
Formula Weight 238.3
Percent Purity ≥99%
Assay Dry basis
Chemical Name or Material HEPES

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.